Learn More
Abcam IACS-010759, 1MG
Supplier:  Abcam AB2869611MG
Encompass_Preferred
MW 562.6 Da, Purity >=98%. A potent and selective Oxidative Phosphorylation Inhibitor (IC₅₀ < 10 nM) with potential antineoplastic activity. IACS-010759 robustly inhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS, likely owing to a combination of energy depletion and reduced aspartate production that leads to impaired nucleotide biosynthesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5804