Learn More
Medchemexpress LLC HY-112037A 50mg , IACS-010759 (hydrochloride) CAS:1807523-99-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112037A50MG
Encompass_Preferred
HY-112037A 50mg IACS-010759 (hydrochloride) CAS: 1807523-99-4 IACS-010759 hydrochlorideis an orally active, potent mitochondrial complex I of oxidative phosphorylation (OXPHOS) inhibitor. IACS-010759 hydrochlorideinhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS. IACS-010759 hydrochloride has the potential for relapsed/refractory AML and solid tumors research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0542