Learn More
Medchemexpress LLC HY-112037A 10mg Medchemexpress, IACS-10759 Hydrochloride CAS:1807523-99-4 Purity:>98%
SDP
Catalog No. 501968993
Encompass_Preferred
Medchemexpress, HY-112037A 10mg IACS-10759 HCl CAS:1807523-99-4 IACS-10759 Hydrochloride is a potent inhibitor of complex I of oxidative phosphorylation (OXPHOS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8993 Supplier Medchemexpress LLC Supplier No. HY112037A10MG
Medchemexpress, HY-112037A 10mg IACS-10759 HCl CAS:1807523-99-4 IACS-10759 Hydrochloride is a potent inhibitor of complex I of oxidative phosphorylation (OXPHOS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.