Learn More
Medchemexpress LLC HY-112037A 5mg Medchemexpress, IACS-10759 Hydrochloride CAS:1807523-99-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112037A5MG
Encompass_Preferred
Medchemexpress, HY-112037A 5mg IACS-10759 Hydrochloride CAS:1807523-99-4 IACS-10759 Hydrochloride is a potent inhibitor of complex I of oxidative phosphorylation ( OXPHOS ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2412