Learn More
Medchemexpress LLC IACS-13909 | 2160546-07-4 | 100.0% | 377.27 | 1 ML
SDP
Catalog No. 5000351096
Encompass_Preferred
IACS-13909 is a selective, potent, and orally active SHP2 allosteric inhibitor with an IC50 of 15.7 nM and a Kd of 32 nM. It demonstrates greater selectivity for SHP2 compared to other phosphatases, including SHP1. This inhibitor exhibits antitumor activities and inhibits MAPK pathway signaling in receptor tyrosine kinase (RTK)-dependent cancers.
  • Selective, potent, and orally active SHP2 allosteric inhibitor
  • IC50 of 15.7 nM and Kd of 32 nM for SHP2
  • Greater selectivity for SHP2 compared to other phosphatases
  • Exhibits antitumor activities
  • Inhibits MAPK pathway signaling in RTK-dependent cancers
Catalog No. 50-003-51096 Supplier Medchemexpress LLC Supplier No. HY13709210MM/1ML
IACS-13909 is a selective, potent, and orally active SHP2 allosteric inhibitor with an IC50 of 15.7 nM and a Kd of 32 nM. It demonstrates greater selectivity for SHP2 compared to other phosphatases, including SHP1. This inhibitor exhibits antitumor activities and inhibits MAPK pathway signaling in receptor tyrosine kinase (RTK)-dependent cancers.
  • Selective, potent, and orally active SHP2 allosteric inhibitor
  • IC50 of 15.7 nM and Kd of 32 nM for SHP2
  • Greater selectivity for SHP2 compared to other phosphatases
  • Exhibits antitumor activities
  • Inhibits MAPK pathway signaling in RTK-dependent cancers