Learn More
Medchemexpress LLC IACS-13909 | 2160546-07-4 | 99.97% | 377.27 | 25 MG
SDP
Catalog No. 5000348052
Encompass_Preferred
IACS-13909 is a selective, potent, and orally active SHP2 allosteric inhibitor with an IC50 of 15.7 nM and a Kd of 32 nM. It demonstrates greater selectivity for SHP2 compared to other phosphatases, including SHP1. This compound exhibits antitumor activities and effectively suppresses MAPK pathway signaling in receptor tyrosine kinase (RTK)-dependent cancers.
  • Selective, potent, and orally active SHP2 allosteric inhibitor
  • Greater selectivity for SHP2 compared to other phosphatases
  • Exhibits antitumor activities
  • Suppresses MAPK pathway signaling in RTK-dependent cancers
Catalog No. 50-003-48052 Supplier Medchemexpress LLC Supplier No. 25MGHY137092
IACS-13909 is a selective, potent, and orally active SHP2 allosteric inhibitor with an IC50 of 15.7 nM and a Kd of 32 nM. It demonstrates greater selectivity for SHP2 compared to other phosphatases, including SHP1. This compound exhibits antitumor activities and effectively suppresses MAPK pathway signaling in receptor tyrosine kinase (RTK)-dependent cancers.
  • Selective, potent, and orally active SHP2 allosteric inhibitor
  • Greater selectivity for SHP2 compared to other phosphatases
  • Exhibits antitumor activities
  • Suppresses MAPK pathway signaling in RTK-dependent cancers