Learn More
Medchemexpress LLC IACS-8803 diammonium | 00-00-0 | 96.7% | C20H29FN12O9P2S2 | 10MG
SDP
Catalog No. 5000260425
Encompass_Preferred
IACS-8803 (diammonium) is a cyclic dinucleotide STING agonist supplied as a diammonium salt for research use. It potently activates the STING pathway and has demonstrated robust systemic antitumor efficacy in preclinical studies. The diammonium salt improves handling and stability compared with the free form and is intended for in vitro and in vivo immuno-oncology research.
  • Potent STING pathway agonist for immune activation.
  • Demonstrated systemic antitumor efficacy in preclinical models.
  • Suitable for in vitro and in vivo research applications.
  • Diammonium salt formulation for improved stability and handling.
  • Requires cold-chain storage; research use only.
Catalog No. 50-002-60425 Supplier Medchemexpress LLC Supplier No. HY130115B10MG
IACS-8803 (diammonium) is a cyclic dinucleotide STING agonist supplied as a diammonium salt for research use. It potently activates the STING pathway and has demonstrated robust systemic antitumor efficacy in preclinical studies. The diammonium salt improves handling and stability compared with the free form and is intended for in vitro and in vivo immuno-oncology research.
  • Potent STING pathway agonist for immune activation.
  • Demonstrated systemic antitumor efficacy in preclinical models.
  • Suitable for in vitro and in vivo research applications.
  • Diammonium salt formulation for improved stability and handling.
  • Requires cold-chain storage; research use only.