Learn More
Medchemexpress LLC Iacs-8803 diammonium | 00-00-0 | 98.8% | 726.60 g/mol | C20H29FN12O9P2S2 | 5 MG
SDP
Catalog No. 5000263010
Encompass_Preferred
IACS-8803 diammonium is a potent cyclic dinucleotide STING agonist for preclinical research that activates the STING pathway and has demonstrated systemic antitumor efficacy. The diammonium salt form improves stability for handling and experimental use and is supplied with supporting analytical and safety documentation for research workflows.
  • High purity suitable for research use (98.8%).
  • Potent STING agonist activity for immuno-oncology studies.
  • Suitable for both in vitro and in vivo preclinical experiments.
  • Provided with SDS, COA, HNMR, LCMS, and analytical reports.
  • Available in multiple small research pack sizes for dose-ranging studies.
Catalog No. 50-002-63010 Supplier Medchemexpress LLC Supplier No. HY130115B5MG
IACS-8803 diammonium is a potent cyclic dinucleotide STING agonist for preclinical research that activates the STING pathway and has demonstrated systemic antitumor efficacy. The diammonium salt form improves stability for handling and experimental use and is supplied with supporting analytical and safety documentation for research workflows.
  • High purity suitable for research use (98.8%).
  • Potent STING agonist activity for immuno-oncology studies.
  • Suitable for both in vitro and in vivo preclinical experiments.
  • Provided with SDS, COA, HNMR, LCMS, and analytical reports.
  • Available in multiple small research pack sizes for dose-ranging studies.