Learn More
Medchemexpress LLC HY-130115A 1mg Medchemexpress, IACS-8803 (disodium) CAS:2243079-36-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY130115A1MG
Encompass_Preferred
Medchemexpress, HY-130115A 1mg IACS-8803 (disodium) CAS:2243079-36-7 IACS-8803 disodium is a highly potent cyclic dinucleotide STING agonist. IACS-8803 disodium has a robust systemic antitumor efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1226