Learn More
Medchemexpress LLC IACS-8803 disodium | 2243079-36-7 | 98.8% | 736.50 g·mol⁻¹ | C20H21FN10Na2O9P2S2 | 5 MG
SDP
Catalog No. 5000262097
Encompass_Preferred
IACS-8803 disodium is a potent cyclic dinucleotide STING agonist provided as the disodium salt for preclinical research. It activates STING signaling to stimulate interferon responses and has demonstrated robust systemic antitumor efficacy in mouse models. The compound is a white to off-white solid with high reported purity and is supplied in small solid quantities or as a DMSO solution.
  • Activates STING pathway in vitro and in vivo.
  • Demonstrates robust systemic antitumor efficacy in preclinical models.
  • High purity suitable for research applications.
  • Soluble in DMSO and moderately soluble in water; vehicle protocols available for in vivo use.
  • Store under nitrogen, away from moisture; follow cold storage recommendations.
  • Available in small-mass solid quantities and as a prepared solution for convenience.
Catalog No. 50-002-62097 Supplier Medchemexpress LLC Supplier No. HY130115A5MG
IACS-8803 disodium is a potent cyclic dinucleotide STING agonist provided as the disodium salt for preclinical research. It activates STING signaling to stimulate interferon responses and has demonstrated robust systemic antitumor efficacy in mouse models. The compound is a white to off-white solid with high reported purity and is supplied in small solid quantities or as a DMSO solution.
  • Activates STING pathway in vitro and in vivo.
  • Demonstrates robust systemic antitumor efficacy in preclinical models.
  • High purity suitable for research applications.
  • Soluble in DMSO and moderately soluble in water; vehicle protocols available for in vivo use.
  • Store under nitrogen, away from moisture; follow cold storage recommendations.
  • Available in small-mass solid quantities and as a prepared solution for convenience.