Learn More
Medchemexpress LLC IACS-8803 disodium | 2243079-36-7 | 99.97% | 25 MG
SDP
Catalog No. 5000338760
Encompass_Preferred
IACS-8803 disodium is a highly potent cyclic dinucleotide STING agonist that has demonstrated robust systemic antitumor efficacy. Studies show it exhibits superior antitumor activity, particularly via intra-tumoral injection.
  • Highly potent cyclic dinucleotide STING agonist
  • Demonstrates robust systemic antitumor efficacy
  • Exhibits superior antitumor activity via intra-tumoral injection
  • Store solid at -20°C under nitrogen, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, under nitrogen, away from moisture
Catalog No. 50-003-38760 Supplier Medchemexpress LLC Supplier No. 25MGHY130115A
IACS-8803 disodium is a highly potent cyclic dinucleotide STING agonist that has demonstrated robust systemic antitumor efficacy. Studies show it exhibits superior antitumor activity, particularly via intra-tumoral injection.
  • Highly potent cyclic dinucleotide STING agonist
  • Demonstrates robust systemic antitumor efficacy
  • Exhibits superior antitumor activity via intra-tumoral injection
  • Store solid at -20°C under nitrogen, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, under nitrogen, away from moisture