Learn More
Medchemexpress LLC Medchemexpress LLC HY-102000B 5mg, IACS-9571 Hydrochloride CAS: 2319611-93-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY102000B5MG
Encompass_Preferred
IACS-9571 (ASIS-P040) hydrochloride is a potent and selective inhibitor of TRIM24 and BRPF1, with an IC50 of 8 nM for TRIM24, and Kds of 31 nM and 14 nM for TRIM24 and BRPF1, respectively.
Catalog No. NC2612474