Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IACS-9571 hydrochloride | 2319611-93-1 | 98.3% | 679.22 | C32H43ClN4O8S | 50 MG
SDP

Catalog No. 5000247401
Encompass_Preferred

IACS-9571 hydrochloride is the hydrochloride salt form of a potent, selective inhibitor of the epigenetic readers TRIM24 and BRPF1. It is supplied as a solid for research use and offers improved aqueous solubility and stability versus the free base.

  • Potent, selective inhibitor of TRIM24 and BRPF1 (IC50 8 nM; Kd 31 nM and 14 nM).
  • High purity suitable for research (98.3%).
  • Molecular weight 679.22; formula C32H43ClN4O8S.
  • Solid, light brown to brown appearance; sealed storage recommended.
  • Supplied with product data sheet and SDS for handling and storage.

Catalog No. 50-002-47401 Supplier Medchemexpress LLC Supplier No. HY102000B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IACS-9571 hydrochloride is the hydrochloride salt form of a potent, selective inhibitor of the epigenetic readers TRIM24 and BRPF1. It is supplied as a solid for research use and offers improved aqueous solubility and stability versus the free base.

  • Potent, selective inhibitor of TRIM24 and BRPF1 (IC50 8 nM; Kd 31 nM and 14 nM).
  • High purity suitable for research (98.3%).
  • Molecular weight 679.22; formula C32H43ClN4O8S.
  • Solid, light brown to brown appearance; sealed storage recommended.
  • Supplied with product data sheet and SDS for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.