Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IACS-9571 hydrochloride | 2319611-93-1 | 98.3% | 679.22 g/mol | C32H43ClN4O8S | 1 ML
SDP

Catalog No. 5000247473
Encompass_Preferred

IACS-9571 hydrochloride is a potent, selective small-molecule inhibitor of the bromodomain proteins TRIM24 and BRPF1. It is used in biochemical and cellular research to probe bromodomain function, assess target engagement, and study downstream epigenetic regulation in vitro and in cells.

  • Potent TRIM24 inhibition (IC50 = 8 nM) and strong BRPF1 binding (Kd = 14 nM).
  • Demonstrates cellular activity (EC50 ≈ 50 nM).
  • High purity suitable for research applications (>98% reported by supplier).
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for custom preparation.
  • Molecular formula C32H43ClN4O8S; molecular weight 679.22 g/mol.

Catalog No. 50-002-47473 Supplier Medchemexpress LLC Supplier No. HY102000B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IACS-9571 hydrochloride is a potent, selective small-molecule inhibitor of the bromodomain proteins TRIM24 and BRPF1. It is used in biochemical and cellular research to probe bromodomain function, assess target engagement, and study downstream epigenetic regulation in vitro and in cells.

  • Potent TRIM24 inhibition (IC50 = 8 nM) and strong BRPF1 binding (Kd = 14 nM).
  • Demonstrates cellular activity (EC50 ≈ 50 nM).
  • High purity suitable for research applications (>98% reported by supplier).
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for custom preparation.
  • Molecular formula C32H43ClN4O8S; molecular weight 679.22 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.