Learn More
Medchemexpress LLC IACS-9571 hydrochloride | 2319611-93-1 | 98.3% | 679.22 g·mol⁻¹ | C32H43ClN4O8S | 100 MG
SDP
Catalog No. 5000247298
Encompass_Preferred
IACS-9571 hydrochloride is a potent, selective small-molecule inhibitor of the TRIM24 and BRPF1 bromodomains, used as a research tool in epigenetics and bromodomain biology. It exhibits nanomolar potency and is supplied as a high-purity research chemical with recommended storage guidance.
  • Potent dual TRIM24 and BRPF1 bromodomain inhibition (IC50 8 nM; Kd 31 nM and 14 nM).
  • High purity suitable for biochemical and cellular assays.
  • Available as solid and DMSO solution formats for flexible dosing.
  • Multiple pack sizes to support screening and follow-up experiments.
  • Recommended sealed storage to maintain stability.
Catalog No. 50-002-47298 Supplier Medchemexpress LLC Supplier No. HY102000B100MG
IACS-9571 hydrochloride is a potent, selective small-molecule inhibitor of the TRIM24 and BRPF1 bromodomains, used as a research tool in epigenetics and bromodomain biology. It exhibits nanomolar potency and is supplied as a high-purity research chemical with recommended storage guidance.
  • Potent dual TRIM24 and BRPF1 bromodomain inhibition (IC50 8 nM; Kd 31 nM and 14 nM).
  • High purity suitable for biochemical and cellular assays.
  • Available as solid and DMSO solution formats for flexible dosing.
  • Multiple pack sizes to support screening and follow-up experiments.
  • Recommended sealed storage to maintain stability.