Learn More
Selleck Chemical LLC Iadademstat
SDP
Catalog No. 5000003393
Encompass_Preferred
ORY-1001 (RG-6016) 2HCl is an orally active and selective lysine-specific demethylase LSD1/KDM1A inhibitor with IC50 of 20 nM with high selectivity against related FAD dependent aminoxidases Phase 1
Catalog No. 50-000-03393 Supplier Selleck Chemical LLC Supplier No. S779525MG
ORY-1001 (RG-6016) 2HCl is an orally active and selective lysine-specific demethylase LSD1/KDM1A inhibitor with IC50 of 20 nM with high selectivity against related FAD dependent aminoxidases Phase 1