Learn More
Aviva Systems Biology Corp IAPP Antibody - N-terminal region (ARP41728_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41728_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IAPP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human IAPP. Alternate names: AMYLIN, DAP, IAPApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit
Catalog No. 50-284-950