Learn More
Aviva Systems Biology Corp IARS Antibody - middle region (ARP54954_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54954_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IARS. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human IARS. Alternate names: FLJ20736, IARS1, ILRS, PRO0785, IRS, ILERSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-982