Learn More
Medchemexpress LLC HY-125171 5mg Medchemexpress, IAXO-102 CAS:1115270-63-7 Purity:>98%
SDP
Catalog No. 501932601
Encompass_Preferred
Medchemexpress, HY-125171 5mg IAXO-102 CAS:1115270-63-7 IAXO-102 is a TLR4 antagonist which negatively regulates TLR4 signalling. It inhibits MAPK and p65 NF-κB phosphorylation and expression of TLR4 dependent proinflammatory protein. IAXO-102 also prevents experimental abdominal aortic aneurysm development [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2601 Supplier Medchemexpress LLC Supplier No. HY1251715MG
Medchemexpress, HY-125171 5mg IAXO-102 CAS:1115270-63-7 IAXO-102 is a TLR4 antagonist which negatively regulates TLR4 signalling. It inhibits MAPK and p65 NF-κB phosphorylation and expression of TLR4 dependent proinflammatory protein. IAXO-102 also prevents experimental abdominal aortic aneurysm development [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.