Learn More
IBI SCIENTIFIC 3P QS-710 Quick Screen Horizontal Electrophoresis System, 7 x 10 cm
Catalog No. NC0887537
Encompass_Preferred
IBIs QS-710 Quick-Screening Electrophoresis Gel Box is designed for rapid, linear separations of nucleic acids. It requires minimal setup and provides excellent band resolution with consistent lane-to-lane mobility for accurate results. The QS-710 is versatile for use with applications requiring quick screens such as fragment analysis and recombinant screening in 30 minutes or less.Gel Dimensions: 7.0 cm (wide) x 10.0 cm (long) Maximum Sample Capacity: 30 samples (2 combs, 15 samples ea);Buffer Capacity: 250 ml; Distance Between Electrodes: 19.3 cm; Maximum Voltage: 200 V;Comes with: vented lid, UVT casting tray, casting fixture, two 1.5 mm x 8 tooth combs, bubble level, and power cords
Catalog No. NC0887537 Supplier IBI SCIENTIFIC 3P Supplier No. IB51000
IBIs QS-710 Quick-Screening Electrophoresis Gel Box is designed for rapid, linear separations of nucleic acids. It requires minimal setup and provides excellent band resolution with consistent lane-to-lane mobility for accurate results. The QS-710 is versatile for use with applications requiring quick screens such as fragment analysis and recombinant screening in 30 minutes or less.Gel Dimensions: 7.0 cm (wide) x 10.0 cm (long) Maximum Sample Capacity: 30 samples (2 combs, 15 samples ea);Buffer Capacity: 250 ml; Distance Between Electrodes: 19.3 cm; Maximum Voltage: 200 V;Comes with: vented lid, UVT casting tray, casting fixture, two 1.5 mm x 8 tooth combs, bubble level, and power cords