Learn More
AdipoGen IBA
SDP
Supplier:  AdipoGen CDXH0103M100
Encompass_Preferred
Chemical. CAS 131-62-4. Formula C7H5IO3. MW 264.1. Synthetic. Heterocyclic iodane with catalytic activity in the hydrolytic cleavage of reactive phosphates, phosphonates and carboxylates. Used as catalyst for the micellar cleavage of activated esters and phosphates and shown to be powerful reagent for the cleavage of phosphates in an aqueous cetyltrimethylammonium chloride CTAC micellar medium.
Catalog No. 50-256-0731