Learn More
Alkali Scientific IBA [Indole 3 butyric Acid], 25 Grams
SDP
Catalog No. 5000661526
Encompass_Preferred
Indole-3-butyric acid (IBA) is a potent plant growth regulator in powder form, widely used as a rooting agent in laboratory experiments to stimulate root development in plant cuttings. IBA promotes adventitious root formation, particularly in woody plants and herbaceous species, by stimulating cell division in the root meristem. It is commonly applied in plant tissue culture and horticultural research to enhance rooting efficiency, especially for hard-to-root species. IBA is a key component in plant propagation, allowing for faster and more successful rooting of cutting material and is widely used in both academic and commercial horticulture applications.
Catalog No. 50-006-61526 Supplier Alkali Scientific Supplier No. BAI001CP
Indole-3-butyric acid (IBA) is a potent plant growth regulator in powder form, widely used as a rooting agent in laboratory experiments to stimulate root development in plant cuttings. IBA promotes adventitious root formation, particularly in woody plants and herbaceous species, by stimulating cell division in the root meristem. It is commonly applied in plant tissue culture and horticultural research to enhance rooting efficiency, especially for hard-to-root species. IBA is a key component in plant propagation, allowing for faster and more successful rooting of cutting material and is widely used in both academic and commercial horticulture applications.