Learn More
Alkali Scientific IBA [Indole 3 butyric Acid], 5 Grams
SDP
Catalog No. 5000660326
Encompass_Preferred
Indole-3-butyric acid (IBA) is a plant hormone used as a rooting stimulant in plant tissue culture. This reagent, available in 5 g amounts, is widely used to promote root development in cuttings and other plant tissues during propagation. IBA is a synthetic auxin, which is crucial for initiating and promoting root formation in plants. It is commonly used in laboratories for rooting experiments, particularly for plant species that have difficulty forming roots naturally. IBA is an essential tool for researchers working on plant propagation, tissue culture optimization, and studies of plant growth hormones. The 5 g package size is ideal for medium to large-scale laboratory applications.
Catalog No. 50-006-60326 Supplier Alkali Scientific Supplier No. BAI002CP
Indole-3-butyric acid (IBA) is a plant hormone used as a rooting stimulant in plant tissue culture. This reagent, available in 5 g amounts, is widely used to promote root development in cuttings and other plant tissues during propagation. IBA is a synthetic auxin, which is crucial for initiating and promoting root formation in plants. It is commonly used in laboratories for rooting experiments, particularly for plant species that have difficulty forming roots naturally. IBA is an essential tool for researchers working on plant propagation, tissue culture optimization, and studies of plant growth hormones. The 5 g package size is ideal for medium to large-scale laboratory applications.