Learn More
ECM Bioscience IκBα Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068729
Encompass_Preferred
The IκBα phospho-regulation antibody sampler kit can be used to detect IκBα phosphorylation on Ser-32/Ser-36, Tyr-42, and Tyr-305. The kit also includes a polyclonal antibody to monitor total IκBα expression levels and secondary reagents for detection of these antibodies. Applications: WB, E, IP; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8729 Supplier ECM Bioscience Supplier No. IK6320
The IκBα phospho-regulation antibody sampler kit can be used to detect IκBα phosphorylation on Ser-32/Ser-36, Tyr-42, and Tyr-305. The kit also includes a polyclonal antibody to monitor total IκBα expression levels and secondary reagents for detection of these antibodies. Applications: WB, E, IP; Species Reactivity: Hu, Rt, Ms