Learn More
Sino Biological IBA1 / AIF1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 14227R031100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IBA1 / AIF1 (rh IBA1 / AIF1; Catalog#14227-H07E; P55008-1; Met1-Pro147).
Catalog No. 50-224-8003