Learn More
Biomatik Corporation IBA1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50168
Encompass_Preferred
IBA1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Allograft Inflammatory Factor 1. The IBA1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 93~147aa. Applications: ELISA, WB, IF, IHC. Target Synonyms: Allograft Inflammatory Factor 1. Accession Number: P55008. Gene ID: 199. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4384