Learn More
ACROBiosystems ACROBiosystems Human Iba1 / AIF-1 Protein, His Tag
Catalog No. 501956407
Encompass_Preferred
Recombinant Protein;1MG;Human Iba1, His Tag (IB1-H51H9) is expressed from E.coli cells. It contains AA Met 1 - Pro 147 (Accession # P55008-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.7 kDa.
Catalog No. 50-195-6407 Supplier ACROBiosystems Supplier No. IB1H51H91MG
Recombinant Protein;1MG;Human Iba1, His Tag (IB1-H51H9) is expressed from E.coli cells. It contains AA Met 1 - Pro 147 (Accession # P55008-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.7 kDa.