Learn More
Cell Signaling Technology Iba1/AIF-1 Monoclonal Antibody, Rabbit, Human and Mouse Reactivity, 25 μl
SDP
Catalog No. 503133858
Encompass
The Iba1/AIF-1 (E4O4W) Rabbit Monoclonal Antibody is a recombinant antibody engineered to detect endogenous levels of total Iba1/AIF-1 protein. Developed by immunizing animals with a synthetic peptide corresponding to residues surrounding Ala139 of the human protein, this antibody is specifically conjugated for use in single cell analysis workflows. It serves as a precise tool for the identification of microglia and macrophages in various tissue samples.
  • Validated for single cell analysis applications
  • Reacts with human and mouse species
  • Supplied in a formulation containing 50% glycerol, PBS, 2 mM EDTA, 0.05% Triton X-100, and 2 mg/mL BSA
  • Developed as a recombinant rabbit monoclonal antibody
  • Recognizes endogenous levels of the total target protein
Catalog No. 50-313-3858 Supplier Cell Signaling Technology Supplier No. 82746S
The Iba1/AIF-1 (E4O4W) Rabbit Monoclonal Antibody is a recombinant antibody engineered to detect endogenous levels of total Iba1/AIF-1 protein. Developed by immunizing animals with a synthetic peptide corresponding to residues surrounding Ala139 of the human protein, this antibody is specifically conjugated for use in single cell analysis workflows. It serves as a precise tool for the identification of microglia and macrophages in various tissue samples.
  • Validated for single cell analysis applications
  • Reacts with human and mouse species
  • Supplied in a formulation containing 50% glycerol, PBS, 2 mM EDTA, 0.05% Triton X-100, and 2 mg/mL BSA
  • Developed as a recombinant rabbit monoclonal antibody
  • Recognizes endogenous levels of the total target protein