Learn More
Selleck Chemical LLC Iberdomide (CC220) 2mg 1323403-33-3
SDP
Supplier:  Selleck Chemical LLC S87602MG
Encompass_Preferred
Iberdomide (CC-220) is a novel oral immunomodulatory compound that targets cereblon, part of the CRL4CRBN E3 ubiquitin ligase complex, with an IC50 of 60 nM in a competitive TR-FRET assay. Iberdomide (CC-220) induces apoptosis with antitumor and immunostimulatory activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8219