Learn More
Lkt Laboratories Iberin, ≥97%, 10MG
SDP
Supplier:  Lkt Laboratories I041610MG
Encompass_Preferred
Isothiocyanate found in cruciferous vegetables, homolog of sulforaphane. CAS: 505-44-2. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4857