Learn More
Medchemexpress LLC Iberiotoxin (TFA) | 4230.85 | 500 UG
SDP
Catalog No. 5000425642
Encompass_Preferred
Iberiotoxin (TFA) is a selective high conductance Ca2+-activated K+ channel inhibitor with a Kd of approximately 1 nM. It does not block other types of voltage-dependent ion channels.
  • Selective high conductance Ca2+-activated K+ channel inhibitor
  • Kd of approximately 1 nM
  • Does not block other types of voltage-dependent ion channels
Catalog No. 50-004-25642 Supplier Medchemexpress LLC Supplier No. HYP0190A500UG
Iberiotoxin (TFA) is a selective high conductance Ca2+-activated K+ channel inhibitor with a Kd of approximately 1 nM. It does not block other types of voltage-dependent ion channels.
  • Selective high conductance Ca2+-activated K+ channel inhibitor
  • Kd of approximately 1 nM
  • Does not block other types of voltage-dependent ion channels