Learn More
IBI SCIENTIFIC 3P Basic Agarose | 9012-36-6 | MFCD00081294 | 100 g
Catalog No. 50550470
Encompass_Preferred
IBI Basic Agarose is a molecular biology certified agarose for use in standard electrophoresis procedures. IBI Basic has excellent gel strength and clarity providing clear concise bands. It is a high-quality, low-cost alternative.
Catalog No. 50-550-470 Supplier IBI SCIENTIFIC 3P Supplier No. IB70070
IBI Basic Agarose is a molecular biology certified agarose for use in standard electrophoresis procedures. IBI Basic has excellent gel strength and clarity providing clear concise bands. It is a high-quality, low-cost alternative.