Learn More
IBI SCIENTIFIC 3P Basic Agarose | 9012-36-6 | MFCD00081294 | 500 g
Catalog No. 50550472
Encompass_Preferred
IBI Basic Agarose - 500gm IBI introduces Basic Agarose! A molecular biology certified agarose for use in standard electrophoresis rocedures. IBI Basic has excellent gel strength and clarity, and provides clear concise bands.
Catalog No. 50-550-472 Supplier IBI SCIENTIFIC 3P Supplier No. IB70072
IBI Basic Agarose - 500gm IBI introduces Basic Agarose! A molecular biology certified agarose for use in standard electrophoresis rocedures. IBI Basic has excellent gel strength and clarity, and provides clear concise bands.