Learn More
IBI SCIENTIFIC 3P MP-1015 Multi Purpose Horizontal Electrophoresis System, 10 x 15 cm
Catalog No. NC9707998
Encompass_Preferred
MP-1015, Multi-Purpose Electrophoresis Gel Box (10x15cm.) General purpose unit for high-resolution linear runs and quick screens. Constructed of cast acrylic. Buffer recirculation. Multi-channel pipette compatible. Max Sample Cap. = 80.
Catalog No. NC9707998 Supplier IBI SCIENTIFIC 3P Supplier No. IB53000
MP-1015, Multi-Purpose Electrophoresis Gel Box (10x15cm.) General purpose unit for high-resolution linear runs and quick screens. Constructed of cast acrylic. Buffer recirculation. Multi-channel pipette compatible. Max Sample Cap. = 80.