Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBI Scientific™ Clear Polycarbonate Culture Medium Bottles

Catalog No. 15453237 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Culture medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
15-453-237 Culture medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-453-237 Supplier IBI Scientific™ Supplier No. ACCFL0008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Flow Cells

  • Clear polycarbonate
  • Autoclavable
  • Contain silicon tubing and fixtures for delivery of medium directly to flow cells
  • Polypropylene cap
  • Available in 4L and 10L

Specifications

Type Culture Medium Bottle
Autoclavable Autoclavable
Capacity (Metric) 4 L
Color Clear
For Use With (Equipment) Culture medium
Includes 12 in. L of 0.12 in. I.D. Silicone tubing and closures for delivery of medium or capture of waste effluent
Material Polycarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.