Learn More
IBI Scientific™ Convertible Flow Cell, Complete System
Catalog No. 15455101 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
Material:
APET Coverslip
Glass Coverslip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
15-455-101 APET Coverslip
15-455-100 Glass Coverslip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-455-101 Supplier IBI Scientific™ Supplier No. CFCAS0002
Add to Cart
Add to Cart
Includes
System Components:
  • Flow cell with attached No. 1 coverslip
  • Three-cylinder acrylic bubble trap
  • Two ABS plastic manifolds (one manifold is attached to the peristaltic influent tubing; the other consolidates the effluent PVC tubing)
Additional Components:
  • 14 in. (35.6cm) length of 0.04 in. (1.1mm) I.D. Tygon™ LFL peristaltic pump tubing
  • 6 ft. (183cm) length total of influent and effluent 1/16 in. (1.5mm) clear PVC USP class VI tubing
  • Luer-Lock™ effluent interrupts
  • Identification tags
  • Six pinch clamps
Gamma-irradiated flow cell apparatus for creation and direct, nondestructive online microscopic examination of bio films
  • Single chamber cell measuring 7.7cm (4mm x 40mm x 8mm)
  • Detachable/reattachable top
  • Tops available in glass (Model CFCAS0001) or plastic cover slip (Model CFCAS0002)
  • Test samples can then be exposed to cultures of interest by injecting through the septum located on the top of the cell frame
  • Full assembly includes all necessary tubing
Order Info
  • Dimensions of each channel: 1 5/8L x 0.2W x 0.04 in.H (40 x 4 x 1mm).
  • Dimensions of coverslip: 2L x 0.09 x 0.006 in.H (50 x 2.4 x 0.2mm).
Specifications
Type Flow Cell Apparatus
Autoclavable Not Autoclavable
Includes Three-cylinder Acrylic bubble trap, 1 removable coverslip, Identification tags, Six pinch clamps
Material APET Coverslip