Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBI Scientific™ Low Profile Roller Shaker

Catalog No. 15453261 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
Voltage:
94/240 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
15-453-261 94/240 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-453-261 Supplier IBI Scientific™ Supplier No. ROLAAUV1S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides convenient fit into laboratory incubators, ovens, and refrigerated units

  • Fits on benchtops, in high humidity and CO2 incubators, high-temperature ovens (to 65°C)
  • Variable speed control: 2 to 38rpm
  • Supplied with six rollers
  • Each roller is electronically gear-driven, without belts or pulleys
  • Rollers are removable for cleaning and to accommodate various-sized tubes or bottles
  • Thin, flat power cord reaches external outlets without damaging incubator gaskets
  • Cool-running 12V DC motor
  • All finishes are chemically resistant, easy to clean and sterilize
  • Aluminum case parts coated in durable, high-bake ester finish
  • Overall dimensions:  38 x 36 x 6cm (15L x 14.25W x 2.37 in. H)
  • Roller dimensions:  2.9 x 25.4cm (1.12 in. dia. x 10 in.L)
  • Net weight: 4.8kg
Applications:
  • Cell culture; Western, Northern and Southern Filter Blots, Antibody Staining

Order Info

Additional roller bottles can be purchased separately

Specifications

Speed Range 2 to 38 rpm
Voltage 94/240 V
Temperature Range (Metric) 0°C to 60°C
Holds 4 L
Height (English) 2.75 in.
Height (Metric) 6.98 cm
For Use With (Application) For rotating cell culture bottles
For Use With (Equipment) Benchtop, High humidity and CO2 incubators, High temperature ovens (60⁰C), and refrigerators (0⁰C)
Humidity Range 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.