Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBI Scientific™ Well Mark™

Catalog No. 15455150 Shop All Stovall Life Science Inc. Products
Change view
Click to view available options
Well Mark system; w/Ergonomic Easel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
15-455-150 Well Mark system; w/Ergonomic Easel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-455-150 Supplier IBI Scientific™ Supplier No. WMKAA0003

missing translation for 'validMainframeMsg'

Used to reduce pipette dispensing errors and cross contamination during material delivery of assay reagents to 96- and 384-Well plates

  • Topas, 8007, HDPE, stainless steel system
  • Windowed frame which locks any skirted or unskirted wellplate in place
  • Moving, protective shield with an open slot revealing one column of wells at a time
  • Single-well isolator with protective skirt that moves down a column, one well at a time
  • Small, hinged acrylic ergonomic easel holds the plate and Well Mark at an angle advantageous for easy deposit of assay reagents
  • Nonskid mats secure plate and Well Mark in place as operator advances the slotted, protective cover from column to succeeding column of wells
  • Easel can be set at three different angles
  • Dimensions: 5 x 7 in. (12.7 x 17.8cm)
  • Easel: 5.5 x 7.5 in. (14 x 19)

Specifications

Diameter (English) Outer 7 in.
Diameter (Metric) Outer 17.78 cm
Dimensions (L x W) 0.75 x 7 x 5 in. LxWxD (1.9 x 17.78 x 12.7 cm)
Material HDPE, Stainless Steel
For Use With (Application) To reduce pipet dispensing errors and cross contamination during material delivery of assay reagents to 96- and 384-well plates
Includes 1 H x 7.5 W x 5.5 in. D Small hinged Acrylic Ergonomic easel and two removable, cleanable, non-skid mats, windowed frame, 384-well plate, operation manual, moving protective shield with an open slot, single-well isolator with protective skirt
Length (English) 0.75 in.
Weight (English) 8 oz.
Weight (Metric) 227 mg
Length (Metric) 1.9 cm
Type Well Mark System
For Use With (Equipment) Opaque, clear and tinted 96- and 384-well microplates
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.