Learn More
Ibidi USA µ-Slide 8 Well high
SDP
Catalog No. NC3036635
Encompass
A chambered coverslip with 8 individual wells, high walls. All-in-one 8 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. Hish-resolution imaging through the #1.5 polymer coverslip bottom with the highest optical quality. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: Poly-L-Lysine: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)
Catalog No. NC3036635 Supplier Ibidi USA Supplier No. 80804
A chambered coverslip with 8 individual wells, high walls. All-in-one 8 well chamber slide for cost-effective experiments with small cell numbers and low reagent volumes. Hish-resolution imaging through the #1.5 polymer coverslip bottom with the highest optical quality. Extra high individual walls to keep cross contamination between wells as low as possible. Surface Modification: Poly-L-Lysine: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)