Learn More
Ibidi USA ibidi Mounting Medium With DAPI, 15ml
SDP
Catalog No. NC1943852
Encompass
DAPI counterstaining and mounting combined in one single step. Ready-to-use in immunofluorescence assays using widefield fluorescence and confocal microscopy. Dropper bottle for exact dispensing into channels or open wells. Brilliant fluorescence signals with low background fluorescence. Concentration/Volume 15mL
This item is not returnable. View return policy
Catalog No. NC1943852 Supplier Ibidi USA Supplier No. 50011
DAPI counterstaining and mounting combined in one single step. Ready-to-use in immunofluorescence assays using widefield fluorescence and confocal microscopy. Dropper bottle for exact dispensing into channels or open wells. Brilliant fluorescence signals with low background fluorescence. Concentration/Volume 15mL