Learn More
Ibidi USA µ-Slide 2 Well Glass Bottom Bulk Pack (90 slides)
SDP
Catalog No. NC3037823
Encompass
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the ¿-Slides and ¿-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The ¿-Slide 2 Well Glass Bottom is a convenient chambered coverslip with 2 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box
Catalog No. NC3037823 Supplier Ibidi USA Supplier No. 8028790
The ibidi product family is comprised of a variety of ¿-Slides and ¿-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The glass bottom versions of the ¿-Slides and ¿-Dishes are especially designed for TIRF, super resolution, and single-molecule applications. The ¿-Slide 2 Well Glass Bottom is a convenient chambered coverslip with 2 wells for cell culture, immunofluorescence, and high-end microscopy. #1.5H D 263 M Schott glass coverslip, uncoated, sterilized, 15 pcs./box