Learn More
Medchemexpress LLC HY-16743 2mg Medchemexpress, Ibiglustat CAS:1401090-53-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY167432MG
Encompass_Preferred
Medchemexpress, HY-16743 2mg Ibiglustat CAS:1401090-53-6 Ibiglustat (Venglustat), a potential therapy for PD Parkinson’s disease, SRT in Fabry’s and Gaucher’s, is a selective, allosteric inhibitor of glucosylceramide synthase (GCS) with ability to cross the blood-brain barrier[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9215