Learn More
Medchemexpress LLC Ibiglustat (L-malic acid) | 1629063-78-0 | MFCD31746910 | 523.57 g/mol | C24H30FN3O7S | 10 MG
SDP
Catalog No. 5000289247
Encompass_Preferred
Ibiglustat (L-malic acid) is the L-malate salt of ibiglustat (venglustat), an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor supplied as a powder for research use. It is intended for in vitro and preclinical studies of lysosomal storage disorders and GBA-associated neurological disease. Key chemical identifiers include CAS 1629063-78-0, formula C24H30FN3O7S, and molecular weight 523.57 g/mol.
  • Powder form suitable for preparation of stock solutions.
  • L-malate salt improves handling and solubility properties.
  • Brain-penetrant GCS inhibitor for CNS disease research.
  • High DMSO solubility (≥ 100 mg/mL) for convenient stock preparation.
  • Stable as powder at -20°C for long-term storage; solvent stability varies by temperature.
  • Chemical identifiers provided for unambiguous compound identification.
Catalog No. 50-002-89247 Supplier Medchemexpress LLC Supplier No. HY16743A10MG
Ibiglustat (L-malic acid) is the L-malate salt of ibiglustat (venglustat), an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor supplied as a powder for research use. It is intended for in vitro and preclinical studies of lysosomal storage disorders and GBA-associated neurological disease. Key chemical identifiers include CAS 1629063-78-0, formula C24H30FN3O7S, and molecular weight 523.57 g/mol.
  • Powder form suitable for preparation of stock solutions.
  • L-malate salt improves handling and solubility properties.
  • Brain-penetrant GCS inhibitor for CNS disease research.
  • High DMSO solubility (≥ 100 mg/mL) for convenient stock preparation.
  • Stable as powder at -20°C for long-term storage; solvent stability varies by temperature.
  • Chemical identifiers provided for unambiguous compound identification.