Learn More
TARGETMOL CHEMICALS INC IBIGLUSTAT L-MALIC ACID 5MG
SDP
Catalog No. 502581519
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ibiglustat (L-Malic acid) (Ibiglustat L-Malic acid) is a selective brain-penetrant and allosteric inhibitor of glucosylceramide synthase. Ibiglustat (L-Malic acid) can be used in studies about PD Parkinson’s disease SRT in Fabry’s and Gaucher’s. purity: 99%
Catalog No. 50-258-1519 Supplier TARGETMOL CHEMICALS INC Supplier No. T115995MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ibiglustat (L-Malic acid) (Ibiglustat L-Malic acid) is a selective brain-penetrant and allosteric inhibitor of glucosylceramide synthase. Ibiglustat (L-Malic acid) can be used in studies about PD Parkinson’s disease SRT in Fabry’s and Gaucher’s. purity: 99%