Learn More
Medchemexpress LLC HY-16743A 5mg Medchemexpress, Ibiglustat (L-Malic acid) CAS:1629063-78-0 Purity:>98%
SDP
Catalog No. 501932779
Encompass_Preferred
Medchemexpress, HY-16743A 5mg Ibiglustat (L-Malic acid) CAS:1629063-78-0 Ibiglustat L-Malic acid (Venglustat L-Malic acid), a potential therapy for PD Parkinson’s disease, SRT in Fabry’s and Gaucher’s, is a selective, allosteric inhibitor of glucosylceramide synthase (GCS) with ability to cross the blood-brain barrier [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2779 Supplier Medchemexpress LLC Supplier No. HY16743A5MG
Medchemexpress, HY-16743A 5mg Ibiglustat (L-Malic acid) CAS:1629063-78-0 Ibiglustat L-Malic acid (Venglustat L-Malic acid), a potential therapy for PD Parkinson’s disease, SRT in Fabry’s and Gaucher’s, is a selective, allosteric inhibitor of glucosylceramide synthase (GCS) with ability to cross the blood-brain barrier [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.