Learn More
Medchemexpress LLC Ibiglustat succinate | 1629063-80-4 | 99.7% | 507.57 | 2 MG
SDP
Catalog No. 5000390926
Encompass_Preferred
Ibiglustat succinate is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. It is used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
  • Orally active
  • Brain-penetrant glucosylceramide synthase (GCS) inhibitor
  • Can prevent additional GL-3 accumulation in Fabry disease cells
Catalog No. 50-003-90926 Supplier Medchemexpress LLC Supplier No. HY16743B2MG
Ibiglustat succinate is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. It is used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
  • Orally active
  • Brain-penetrant glucosylceramide synthase (GCS) inhibitor
  • Can prevent additional GL-3 accumulation in Fabry disease cells