Learn More
Medchemexpress LLC Ibiglustat (succinate) | 1629063-80-4 | 99.7% | 507.57 | 25 MG
SDP
Catalog No. 5000391353
Encompass_Preferred
Ibiglustat succinate is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. It can be used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
  • Orally active
  • Brain-penetrant glucosylceramide synthase (GCS) inhibitor
  • Can prevent additional GL-3 accumulation in Fabry disease cells
Catalog No. 50-003-91353 Supplier Medchemexpress LLC Supplier No. 25MGHY16743B
Ibiglustat succinate is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. It can be used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
  • Orally active
  • Brain-penetrant glucosylceramide synthase (GCS) inhibitor
  • Can prevent additional GL-3 accumulation in Fabry disease cells