Learn More
Medchemexpress LLC Ibiglustat (succinate) | 1629063-80-4 | 99.7% | 507.57 | 50 MG
SDP
Catalog No. 5000390777
Encompass_Preferred
Ibiglustat (Venglustat) succinate is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. It can be utilized for research related to Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
  • Orally active.
  • Brain-penetrant glucosylceramide synthase (GCS) inhibitor.
  • Can be used for research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
Catalog No. 50-003-90777 Supplier Medchemexpress LLC Supplier No. HY16743B50MG
Ibiglustat (Venglustat) succinate is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. It can be utilized for research related to Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
  • Orally active.
  • Brain-penetrant glucosylceramide synthase (GCS) inhibitor.
  • Can be used for research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.