Learn More
Selleck Chemical LLC Ibiglustat succinate-E0646-5MG
SDP
Catalog No. 503137254
Encompass_Preferred
Ibiglustat succinate (Venglustat succinate) is a glucosylceramide (GlcCer) synthase inhibitor that blocks the formation of glucosylceramide
Catalog No. 50-313-7254 Supplier Selleck Chemical LLC Supplier No. E06465MG
Ibiglustat succinate (Venglustat succinate) is a glucosylceramide (GlcCer) synthase inhibitor that blocks the formation of glucosylceramide