Learn More
Ibidi USA ibiSeal 76x114
SDP
Catalog No. NC2941875
Encompass
A self-adhesive cover film for sealing open wells and reservoirs in cell culture and microscopy applications. Convenient sealing of ibidi labware products. Easy to apply – easy to remove. Broad range of applications (e.g., reduction of evaporation, maintenance of sterility, and sample transport)
Catalog No. NC2941875 Supplier Ibidi USA Supplier No. 10874
Add to Cart
Add to Cart
A self-adhesive cover film for sealing open wells and reservoirs in cell culture and microscopy applications. Convenient sealing of ibidi labware products. Easy to apply – easy to remove. Broad range of applications (e.g., reduction of evaporation, maintenance of sterility, and sample transport)